missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate, dibasic dihydrate, 98+%, extra pure, Thermo Scientific Chemicals
42.80 € - 144.00 €
Chemical Identifiers
CAS | 7789-77-7 |
---|---|
Molecular Formula | CaHO4P·2H2O |
Molecular Weight (g/mol) | 172.09 |
MDL Number | MFCD00149621 |
InChI Key | XAAHAAMILDNBPS-UHFFFAOYSA-L |
Synonym | calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen |
PubChem CID | 104805 |
ChEBI | CHEBI:4496 |
IUPAC Name | calcium;hydrogen phosphate;dihydrate |
SMILES | O.O.OP(=O)([O-])[O-].[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10549423
|
Thermo Scientific Acros
389830010 |
1 kg | Plastic bottle |
42.80 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10330584
|
Thermo Scientific
389830050 |
5 kg | Plastic drum |
144.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-77-7 | |
172.09 | |
XAAHAAMILDNBPS-UHFFFAOYSA-L | |
104805 | |
calcium;hydrogen phosphate;dihydrate |
CaHO4P·2H2O | |
MFCD00149621 | |
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen | |
CHEBI:4496 | |
O.O.OP(=O)([O-])[O-].[Ca+2] |
Specifications
7789-77-7 | |
Powder | |
CaHO4P·2H2O | |
MFCD00149621 | |
Solubility in water: 0.2g/L (25°C) | |
O.O.OP(=O)([O-])[O-].[Ca+2] | |
172.09 | |
CHEBI:4496 | |
≥98% | |
Calcium: positive, Phosphate: positive | |
Calcium phosphate, dibasic dihydrate |
White | |
98.0 to 105% (on dried substance) (Complexometry) | |
CaHPO4·2H2O | |
calcium hydrogenphosphate dihydrate, calcium hydrogen phosphate dihydrate, unii-o7tsz97gep, calcium monohydrogen phosphate dihydrate, o7tsz97gep, calcium phosphate dibasic dihydrate, dibasic calcium phosphate dihydrate, phosphoric acid, calcium salt 1:1 , dihydrate, brushite, aksepillen | |
XAAHAAMILDNBPS-UHFFFAOYSA-L | |
calcium;hydrogen phosphate;dihydrate | |
104805 | |
172.09 | |
Extra Pure | |
24.5% to 26.5% (800°C) |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium phosphate, dibasic dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only