missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium phosphate, monobasic, 90%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 7758-23-8 |
Quantity | 1 kg |
Assay Percent Range | 88% min. (Complexometry) |
Molecular Formula | CaH4O8P2 |
Linear Formula | Ca(H2PO4)2 |
MDL Number | MFCD00010898 |
Merck Index | 15,1695 |
Synonym | calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate |
Solubility Information | Solubility in water: insoluble |
InChI Key | YYRMJZQKEFZXMX-UHFFFAOYSA-L |
Show More |
Safety and Handling
Product Identifier
- Calcium phosphate, monobasic
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'