missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals

58.30 € - 285.00 €

Chemical Identifiers

CAS 12167-74-7
Molecular Formula Ca3O8P2
Molecular Weight (g/mol) 310.17
MDL Number MFCD00010904
InChI Key QORWJWZARLRLPR-UHFFFAOYSA-H
Synonym durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
PubChem CID 14781
IUPAC Name tricalcium diphosphate
SMILES [Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036731.36
500 g
58.30 €
500g
Please to purchase this item. Need a web account? Register with us today!
11344257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036731.A3
2 kg
105.00 €
2kg
Please to purchase this item. Need a web account? Register with us today!
11354257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036731.A9
10 kg
285.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12167-74-7
310.17
QORWJWZARLRLPR-UHFFFAOYSA-H
14781
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Ca3O8P2
MFCD00010904
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
tricalcium diphosphate

Specifications

12167-74-7
Powder
Ca3O8P2
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium diphosphate
14781
White powder
Calcium phosphate tribasic, tribasic
White
34-40% Ca
MFCD00010904
Partially soluble in water.
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
310.17
1004.67
3.14g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 235-330-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only