Learn More
Calcium shot, 1cm (0.4 in.) & down, 99% (metals basis), Thermo Scientific™
49.00 € - 460.00 €
Chemical Identifiers
CAS | 7440-70-2 |
---|---|
Molecular Formula | Ca |
Molecular Weight (g/mol) | 40.08 |
MDL Number | MFCD00010897 MFCD00085314 |
InChI Key | OYPRJOBELJOOCE-UHFFFAOYSA-N |
Synonym | unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate |
PubChem CID | 5460341 |
ChEBI | CHEBI:29320 |
IUPAC Name | calcium |
SMILES | [Ca] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-70-2 | |
40.08 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
5460341 | |
calcium |
Ca | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
CHEBI:29320 | |
[Ca] |
Specifications
7440-70-2 | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
Odorless | |
Shot | |
Calcium shot |
Ca | |
UN1401 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
99% | |
1cm (0.4 in.) & down | |
(metals basis) |
Safety and Handling
GHS H Statement
H261
In contact with water releases flammable gas.
P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H261
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM
missing translation for 'einecsNumber' : 213-179-5
missing translation for 'rtecsNumber' : EV8040000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium shot
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P232-Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.