missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium shot, 99.5% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Color | Silver-White |
Appearance | Silvery-white shot |
Assay Percent Range | 99.5% (Metals basis) |
Quantity | 5 g |
Odor | Odorless |
Vapor Pressure | 0 hPa |
Form | Shot |
Chemical Name or Material | Calcium |
Safety and Handling

Product Identifier
- Calcium shot
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- P232-Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium shot, 99.5% (metals basis), Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction