missing translation for 'onlineSavingsMsg'
Learn More

Calcium solution for ICP 10g/L HNO3 matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

173.00 € - 235.00 €

Specifications

Concentration 10±0.02g/L
Packaging Bottle
Type Mass Spectrometry Standard
Analyte or Component Names Ca
Solvent or Matrix HNO3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12986264
View Documents
Fisher Chemical
S 209 250
250 mL Bottle
173.00 €
250mL
Please to purchase this item. Need a web account? Register with us today!
12996264
View Documents
Fisher Chemical
S 209 500
500 mL Bottle
235.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

10±0.02g/L
Mass Spectrometry Standard
HNO3
Bottle
Ca
Videos
Safety and Handling

Safety and Handling

H225; H304; H315; H336; H410

P210; P233; P240; P241; P261; P264; P271; P273; P280; P301+P310; P302+P352; P303+P361+P353; P304+P340; P312; P321; P331; P332+P313; P370+P378; P391; P403+P233; P403 +P235; P405; P501

Danger

missing translation for 'shelfLife' : 12 Months after opening

Product Identifier
  • Single-element Standard Solution for ICP. Calcium (Ca) 10000mg/l in HNO3 5%

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications