missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
51.20 € - 107.00 €
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12942095
|
Fisher Chemical
A 009 100 |
100 mL | Bottle |
51.20 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12962095
|
Fisher Chemical
A 009 500 |
500 mL | Bottle |
107.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
1g/L | |
Single-component Standard Solution | |
MFCD00011349 | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
Liquid |
7697-37-2 | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
H2O | |
Calcium |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)