missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep PLCA2-2Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ca | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Calcium Standard | |
Triple-leached LDPE bottle | |
125 mL | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010897 MFCD00085314 | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLCA2-2Y, -2T, -2X 10,000 PLCA2-3Y,-3X 2-5% HCl 1000μg/mL PLCA1-2X |
7440-70-2, 7697-37-2, 7732-18-5 | |
1.0, 5.0, 94.0 | |
1.0, 5.0, 94.0 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ca | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
Assurance | |
2% HNO3 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, calciumcarbonate
Product intended for laboratory use only