missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

28.35 € - 50.62 €

Chemical Identifiers

MDL Number 10899
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10486801
View Documents
Fisher Chemical
J/8010/05
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10060230
View Documents
Fisher Chemical
J/8010/08
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Calcium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M

Specifications

Chemical Identifiers

10899

Specifications

Colorless
Liquid
HDPE plastic bottle
1
10899
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium solution 1000 ppm

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.