Learn More
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, Thermo Scientific Chemicals
22.31 € - 44.72 €
Specifications
Chemical Name or Material | Calcium standard solution |
---|---|
Concentration | 997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C) |
Name Note | 1 mg/ml Ca in 0.5N HNO3 |
CAS | 7440-70-2, 7697-37-2, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10296390
|
Thermo Scientific Acros
195881000 |
100 mL | Plastic bottle |
22.31 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328050
|
Thermo Scientific Acros
195885000 |
500 mL | Plastic bottle |
44.72 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Calcium standard solution | |
1 mg/ml Ca in 0.5N HNO3 | |
Plastic bottle | |
MFCD00010897 MFCD00085314 | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
AAS | |
Solution |
997 to 1003 μg/mL of Ca(2+) in 0.5N HNO3 (20°C) | |
7440-70-2, 7697-37-2, 7732-18-5 | |
Ca | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
40.08 | |
Soluble in water |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Calcium atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.