missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium stearate, 5.9 to 7.1% (Ca), powder, Thermo Scientific Chemicals
57.59 € - 223.11 €
Chemical Identifiers
CAS | 1592-23-0 |
---|---|
Molecular Formula | C36H70CaO4 |
Molecular Weight (g/mol) | 607.02 |
MDL Number | MFCD00036390 |
InChI Key | CJZGTCYPCWQAJB-UHFFFAOYSA-L |
Synonym | calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates |
PubChem CID | 15324 |
IUPAC Name | calcium;octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13585040
|
Thermo Scientific Acros
449720010 |
1 kg |
57.59 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13595040
|
Thermo Scientific Acros
449720050 |
5 kg |
223.11 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1592-23-0 | |
607.02 | |
CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
15324 | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2] |
C36H70CaO4 | |
MFCD00036390 | |
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates | |
calcium;octadecanoate |
Specifications
145°C to 160°C | |
Powder | |
MFCD00036390 | |
calcium stearate, calcium octadecanoate, flexichem, calcium distearate, octadecanoic acid, calcium salt, aquacal, calstar, flexichem cs, stavinor 30, stearates | |
CJZGTCYPCWQAJB-UHFFFAOYSA-L | |
calcium;octadecanoate | |
15324 | |
6.4 to 7.4% (Ca) | |
Calcium stearate, 6.4 to 7.4% (Ca) |
1592-23-0 | |
C36H70CaO4 | |
15,1705 | |
Solubility in water: insoluble | |
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Ca+2] | |
607.02 | |
607.02 | |
Plastic Bottle |
Safety and Handling
missing translation for 'einecsNumber' : 216-472-8
Product Identifier
- Calcium stearate
RUO – Research Use Only