missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Calcium sulfate, anhydrous is used as adhesives and sealant chemical, Fillers, Fuels and fuel additives, Intermediates, Process regulators, Agricultural Products, Building/Construction Materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1450°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Linear Formula | CaSO4 |
Merck Index | 14,1706 |
Solubility Information | Soluble in water,ammonium salts,sodium thiosulfate & glycerin. |
Formula Weight | 136.14 |
Percent Purity | 99.993% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'