missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Anhydrous, Technical, Granular, Desiccating Agent, Fisher Chemical

Product Code. 10223710
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 1450°C
CAS 7778-18-9
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula CaO4S
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES [Ca++].[O-]S([O-])(=O)=O
IUPAC Name calcium sulfate
Molecular Weight (g/mol) 136.13
Formula Weight 136.14g/mol
Packaging Plastic powder jar
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'