missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate dihydrate, 98+%, extra pure, Thermo Scientific Chemicals
65.50 € - 205.00 €
Chemical Identifiers
CAS | 10101-41-4 |
---|---|
Molecular Formula | CaH4O6S |
Molecular Weight (g/mol) | 172.16 |
MDL Number | MFCD00149625 |
InChI Key | PASHVRUKOFIRIK-UHFFFAOYSA-L |
Synonym | calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone |
PubChem CID | 24928 |
ChEBI | CHEBI:32583 |
IUPAC Name | calcium;sulfate;dihydrate |
SMILES | O.O.[Ca++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12045390
|
Thermo Scientific Acros
446380010 |
1 kg | Plastic bottle |
65.50 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12055390
|
Thermo Scientific Acros
446380050 |
5 kg | Plastic bottle |
205.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-41-4 | |
172.16 | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
24928 | |
calcium;sulfate;dihydrate |
CaH4O6S | |
MFCD00149625 | |
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
CHEBI:32583 | |
O.O.[Ca++].[O-]S([O-])(=O)=O |
Specifications
10101-41-4 | |
CaH4O6S | |
MFCD00149625 | |
Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents | |
O.O.[Ca++].[O-]S([O-])(=O)=O | |
172.16 | |
CHEBI:32583 | |
≥98% | |
Calcium sulfate dihydrate |
98% min. (Complexometry) | |
CaSO4·2H2O | |
calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone | |
PASHVRUKOFIRIK-UHFFFAOYSA-L | |
calcium;sulfate;dihydrate | |
24928 | |
172.17 | |
Extra Pure |
Safety and Handling
RUO – Research Use Only