missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate Dihydrate, ≥99%, ReagentPlus™ Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Color | Colorless |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | CaSO4 · 2H2O |
UN Number | NONH for all modes of transport |
IUPAC Name | calcium;sulfate;dihydrate |
Formula Weight | 172.17g/mol |
Percent Purity | ≥99% |
Odor | Odorless |
Packaging | Poly bottle |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction