missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate dihydrate, 99%, Thermo Scientific Chemicals

Product Code. 11357618
Click to view available options
Quantity:
1 kg
5 kg

Used as portland-cement retarder, tile and plaster. It is a source of sulfur and sulfuric acid. Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium;sulfate;dihydrate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 163°C (in - 2H2O)
Melting Point 128°C (in -1.5H2O)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99%
Merck Index 14,1706
Solubility Information Soluble in water.
Formula Weight 172.17 (136.14 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32 g/mL
Chemical Name or Material Calcium sulfate dihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.