missing translation for 'onlineSavingsMsg'
Learn More

Calcium Sulfate Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 11468972
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10101-41-4
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number 149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color White
pH 7
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 172.17g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'