missing translation for 'onlineSavingsMsg'
Learn More

Calcium sulfate hemihydrate, 97+%, pure

Product Code. p-3523387 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10298653

Brand: Thermo Scientific Acros 385351000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 10034-76-1 | 0·5 H2O | 145.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-76-1
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 145.15
MDL Number MFCD00149624
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
PubChem CID 3033839
ChEBI CHEBI:32584
IUPAC Name dicalcium;disulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 97% min. (On dry substance) (Complexometry)
Solubility Information Solubility in water: 0.8g/L (20°C)
Formula Weight 145.15
Percent Purity ≥97%
Grade Pure
Identification Confirmed
Loss on Ignition 4.5% to 8.0% (500°C, 2 Hrs)
Packaging Plastic bottle
Chemical Name or Material Calcium sulfate hemihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.