missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, Technical, Anhydrous, Powder, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Calcium Sulfate, Technical, Anhydrous, Powder, SLR
Specifications
Specifications
Melting Point | 1450°C |
Color | White |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 136.14g/mol |
Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction