Learn More
Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm, Thermo Scientific Chemicals
Description
It is used as luminous paint, depilatory, lubricant additive, ore dressing and flotation agent and as an arsenic-free hydrogen sulfide. It is also used as a depilatory in the tanning industry and in cosmetics, in homeopathy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | (decomposition) |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN3134 |
Merck Index | 14,1707 |
Solubility Information | Slightly soluble in water. Insoluble in alcohol |
Formula Weight | 72.14 |
Percent Purity | 99.9% |
Odor | Stench |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.