Learn More
Calcium tetrafluoroborate hydrate, Thermo Scientific Chemicals
176.88 € - 526.62 €
Chemical Identifiers
CAS | 13814-93-2 |
---|---|
Molecular Formula | B2CaF8 |
Molecular Weight (g/mol) | 213.685 |
MDL Number | MFCD00015972 |
InChI Key | YHLNCCJTKRTQEG-UHFFFAOYSA-N |
Synonym | calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f |
PubChem CID | 6096284 |
IUPAC Name | calcium;ditetrafluoroborate |
SMILES | [B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312878
|
Thermo Scientific Alfa Aesar
030131.14 |
25 g |
176.88 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302878
|
Thermo Scientific Alfa Aesar
030131.22 |
100 g |
526.62 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium tetrafluoroborate hydrate is used as pharmaceutical and chemical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13814-93-2 | |
213.685 | |
YHLNCCJTKRTQEG-UHFFFAOYSA-N | |
6096284 | |
[B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2] |
B2CaF8 | |
MFCD00015972 | |
calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f | |
calcium;ditetrafluoroborate |
Specifications
13814-93-2 | |
B2CaF8 | |
calcium tetrafluoroborate, calcium ditetrafluoroborate, calcium tetrafluoroborate 1-, acmc-1bo7f | |
YHLNCCJTKRTQEG-UHFFFAOYSA-N | |
calcium;ditetrafluoroborate | |
6096284 | |
Hygroscopic |
Powder | |
MFCD00015972 | |
Soluble in water. | |
[B-](F)(F)(F)F.[B-](F)(F)(F)F.[Ca+2] | |
213.685 | |
213.68 (Anhydrous) | |
Calcium tetrafluoroborate hydrate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 237-485-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.