missing translation for 'onlineSavingsMsg'
Learn More

Calcium trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals

Código de producto. 11369277
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

Calcium trifluoromethanesulfonate is used for aminolysis of epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 55120-75-7
Molecular Formula C2CaF6O6S2
Molecular Weight (g/mol) 338.204
MDL Number MFCD00143911
InChI Key PUQLFUHLKNBKQQ-UHFFFAOYSA-L
Synonym calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate
PubChem CID 2734307
IUPAC Name calcium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]

Especificaciones

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula Ca(OSO2CF3)2
Solubility Information Soluble in water.
Formula Weight 338.21
Percent Purity ≤99%
Chemical Name or Material Calcium trifluoromethanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'