Learn More
Calcium turnings, 98% (metals basis), Thermo Scientific Chemicals
33.57 € - 180.90 €
Chemical Identifiers
CAS | 7440-70-2 |
---|---|
Molecular Formula | Ca |
Molecular Weight (g/mol) | 40.08 |
MDL Number | MFCD00010897 MFCD00085314 |
InChI Key | OYPRJOBELJOOCE-UHFFFAOYSA-N |
Synonym | unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate |
PubChem CID | 5460341 |
ChEBI | CHEBI:29320 |
IUPAC Name | calcium |
SMILES | [Ca] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381168
|
Thermo Scientific Alfa Aesar
010125.09 |
10 g |
33.57 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391168
|
Thermo Scientific Alfa Aesar
010125.22 |
100 g |
180.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-70-2 | |
40.08 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
5460341 | |
calcium |
Ca | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
CHEBI:29320 | |
[Ca] |
Specifications
7440-70-2 | |
98% (metals basis) | |
MFCD00010897 MFCD00085314 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
40.078 | |
Calcium |
1.54 | |
Ca | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
Turnings |
Safety and Handling
P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H261
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: II
missing translation for 'einecsNumber' : 213-179-5
missing translation for 'rtecsNumber' : EV8040000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium turnings
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P332-If skin irritation occurs:.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P233-Keep container tightly closed.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
- P501-Dispose of contents/container to .
RUO – Research Use Only