Learn More
Novex™ Calyculin A
Calculin A
Brand:  Novex™ PHZ1044
Product Code. 10079362
  • 363.00 € / Each
Explore more special offers
Description
Description
Calyculin A is a structurally unique marine toxin with potent and specific protein phosphatase inhibiting activity. This compound inhibits PP2A with similar potency to okadaic acid, but inhibits PP1 with a 10- to 100-fold greater potency.
Format/Formulation: Lyophilized
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5 to -30°C).
5519
TLC
PP2A and PP1 Phosphatases
Phosphatase Inhibitor
1009.2 Da
Dry Ice
Non-sterile
Chemical