missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcyphosine Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16926-1-AP-20UL
This item is not returnable.
View return policy
Specifications
Calcyphosine | |
Polyclonal | |
Unconjugated | |
CAPS | |
Calcyphosin, calcyphosine, CAPS, CAPS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
828 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13938 | |
CAPS | |
Calcyphosine Fusion Protein Ag10330 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |