missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Calibrated Replacement Probes for Traceable™ Data Logging Ethernet Thermometers

Replacement probes that allow you to save time and money by simply replacing probes that are damaged or due for calibration.

Brand:  Fisherbrand™ 61010-98

 View more versions of this product

Product Code. 16556820

  • 200.00 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • No need to send in your thermometer for re-calibration – simply replace the recalibrated probe
  • Get accurate, reliable measurements with included NIST-traceable calibration
Specifications

Specifications

NIST
Reusable
120 in.
304.8 cm
Stainless Steel
Replacement Stainless Steel Probe with Calibration
6601FS Thermometer
12.7 x 7.62 x 22.86 cm (5 x 3 x 9 in.)
Temperature measurement
125 in.
317.5 cm
Thermometer Probe
Yes
±0.10°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.