Learn More
BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
Brand:  BINDER™ 8012-1582
 View more versions of this product
Product Code. 15963002
  • 1345.00 € / Each
Explore more special offers
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
BINDER Chambers (Models MKT 115, MKT 240, MKT 720)
15 to 18 Measurement Points
Temperature Measurement
Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature)
Calibration Certificate (Temperature)