Learn More
Draeger Calibration Gas, H2S, CO, CO2, CH4, O2/N2
Test gas in disposable bottles. Control valve not supplied (please enquire).
Brand:  Draeger 6811132
Product Code. 15330448
  • 505.00 € / Each
Explore more special offers
Description
Description
  • 58L disposable bottle
  • According to ISO 9002.DOT39 and EN 12205
Specifications
Specifications
58 L
2.5 Vol.% / 15 ppm / 50 ppm / 2 Vol.%
Hydrogen Sulfide / Carbon Monoxide / Carbon Dioxide
ISO 9002.DOT39 and EN 12205
35 bar
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Draeger Calibration Gas, H2S, CO, CO2, CH4, O2/N2 >