Learn More
BINDER™ Calibration Service 1 P Pressure with Certificate, Extension for Series VD, VDL
BINDER™ Calibration Service 1 P Pressure Service for use with vacuum drying chambers VD and VDL.
Brand:  BINDER™ DL30-0411
Product Code. 17393703
  • 110.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Calibration service including certificate, measurement at specified pressure
  • Extension of the calibration of the pressure display by another pressure value
  • Including certificate
Specifications
Specifications
Calibration digital pressure display - including certificate, measurement at specified pressure, range 20 - 900 Mbar
Calibration Certificate Expansion
20 to 900 Mbar
BINDER Chambers Series VD, VDL
Digital