Learn More
Agilent BioTek Calibration Solution with 1 Cuvette
Calibration Solution with 1 Cuvette includes the Take3 Calibration Solution and one quartz, stoppered cuvette.
Brand:  Agilent BioTek 4690008
Product Code. 16523042
  • 276.00 € / Each
Explore more special offers
Specifications
Gen5v1.0 and Take3 Module Only, Agilent BioTek Microplate Washer and Reader Instruments
Take3 Calibration Solution (PN: 4693002); 1 quartz cuvette with stopper (PN: 48723)