missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Calmagite, Indicator Grade, Pure Product Code.: 10458440
Add to basket

Thermo Scientific Chemicals Calmagite, Indicator Grade, Pure

Product Code. 10458440
5 g, Glass bottle
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10458440

Brand: Thermo Scientific Chemicals 108170050

Add to basket
Add to basket
Request Bulk or Custom Format
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3147-14-6
Molecular Formula C17H14N2O5S
Molecular Weight (g/mol) 358.368
MDL Number MFCD00011656
InChI Key ASFVMSDYPYMUFL-UHFFFAOYSA-N
Synonym 3-Hydroxy-4-(2-hydroxy-5-methylphenylazo)-1-naphthalenesulfonic acid
PubChem CID 6364506
IUPAC Name 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid
SMILES CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O

Specifications

Melting Point 300.0°C
Merck Index 15, 1720
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 358.38
Grade Pure
Packaging Glass bottle
Specific Extinction 300 min. (610nm, in buffer)
Suitability suitable for determination of calcium and magnesium
Color Black
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Calmagite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463881 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number108170050,108171000,108170250,108170100
Lot NumberA0441755 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number108170050,108171000,108170250,108170100
Lot NumberA0412800 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number108170050,108171000,108170250,108170100
Lot NumberA0448205 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number108170050,108171000,108170250,108170100
Lot NumberA0395201 Certificate TypeCertificate of Analysis Date07/04/2022 Catalog Number108170050,108171000,108170250,108170100
compliance-icons
Product Identifier
  • Calmagite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Calmagite, Indicator Grade, Pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.