missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Calmodulin Sepharose™ 4B Medium

Use for purification of proteins with affinity for calmodulin, a highly conserved regulatory protein involved in many cellular processes in eukaryotic cells. Calmodulin Sepharose™ 4B Medium provides single-step purification.

Brand:  Cytiva 17-0529-01


Product Code. 11504925

  • 689.00 € / 10mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for tandem affinity purification (TAP) of protein complexes
  • Purification of calmodulin-regulated proteins from all eukaryotic cells
  • For single-step purification of native calmodulin-binding proteins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.9 to 1.3mg Calmodulin/mL
4% Agarose
4 to 9
2°C to 8°C
Calmodulin
Cyanogen bromide activation
90 μm
10 mL
For purification of proteins with affinity for calmodulin, a highly conserved regulatory Protein involved in many cellular processes in eukaryotic cells
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calmodulin Sepharose 4B

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers