missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11477-1-AP-150UL
This item is not returnable.
View return policy
Description
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP8, contains two EF-hand domains for calcium binding and shares 70% homology with CaBP7 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. Both CaBP8 and CaBP7 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.Specifications
CALN1 | |
Polyclonal | |
Unconjugated | |
CALN1 | |
CaBP8, Calcium binding protein 8, CALN1, calneuron 1, Calneuron I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140904, 363909, 83698 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q06BI3, Q9BXU9, Q9JJG7 | |
Caln1 | |
CALN1 Fusion Protein Ag2043 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |