missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calorimeter Cover
28.95 € - 29.75 €
Specifications
Product Type | Calorimeter Cover |
---|---|
For Use With (Equipment) | Calorimeter Wood |
Quantity | 1/Ea. |
Product Code | Brand | Product Type | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Width (Metric) | Price | Quantity & Availability | |||||
13136310
|
S Murray SAMCO
Y338/01 |
Calorimeter Cover | 50 mm |
28.95 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
|||||
13146310
|
S Murray SAMCO
Y338/02 |
Calorimeter Cover | 65 mm |
29.75 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
Specifications
Calorimeter Cover | |
Calorimeter Wood | |
1/Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calorimeter Cover