Learn More
Thermo Scientific Chemicals Calphostin C, 95%
Brand: Thermo Scientific Chemicals J60647.!G
This item is not returnable.
View return policy
Description
Potent inhibitor of protein kinase C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Powder | |
790.76 | |
100 μg | |
UCN-1028c |
95% | |
Potent inhibitor of protein kinase C | |
Calphostin C, 95% | |
RUO - research use only |