missing translation for 'onlineSavingsMsg'
Learn More

Honeywell KCL™ Camapur™ Comfort 619 Gloves

Product Code. 15867661
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Honeywell KCL™ Camapur™ Comfort 619 Gloves feature excellent touch sensitivity and high mechanical resistance.

  • Elegant fit and shape
  • Excellent touch sensitivity
  • High mechanical resistance
  • Very good grip even on wet and oily objects
  • Washable
  • Breathable
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EC Category PPE 2
Coated Area Palm
Coating Material Polyurethane
Color White, Gray
Cut Resistance Rating 1
Size 9
Thickness (Metric) 1.0 mm
For Use With (Application) General purpose
Liner Material Polyamide
Puncture Rating 1
Tear Rating 3
Length (Metric) 220 to 260 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'