Learn More
Zeiss™ Camera Adapter P95-C 1/2 in. 0.5x
C mount camera adapter for Primostar 3 - 0.5x
Brand:  Zeiss™ 415501-1811-000
Product Code. 17190676
  • 394.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Requires Primostar 3
  • With phototube C mount camera adapter
  • For Primostar 3 - 0.5x
Specifications
Specifications
Primostar 3 - 0.5 x