Learn More
Motic™ 2.5X Photo Eyepiece for BA/AE Series Trinocular Microscopes
Must be used with Photo Adapter. Motic™ 2.5X Photo Eyepiece mounts to the Reflex camera to a BA/AE trinocular microscope.
Brand:  Motic™ 1101000300442
Product Code. 15129404
  • 129.00 € / 1 unit
Estimated Shipment: 13-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with BA/AE Series Trinocular Microscopes
  • Magnification: 2.5X
  • Requires photo adapter
Order Info
dimProductType
Specifications
Specifications
Camera Occulary
Motic AE2000, AE20/21 or the BA Series Microscopes
2.5X
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ 2.5X Photo Eyepiece for BA/AE Series Trinocular Microscopes > Product Type: Camera Occulary; For Use With Motic AE2000, AE20/21 or the BA Series Microscopes