missing translation for 'onlineSavingsMsg'
Learn More

cAMPS-Sp, triethylammonium salt, Tocris Bioscience™

Product Code. 30001236
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-permeable cAMP analog

Cell-permeable cAMP analog that activates cAMP receptor proteins such as PKA and cAMP-regulated guanine nucleotide exchange factor.

R-enantiomer also available.

Specifications

CAS 93602-66-5
Molecular Formula C16H27N6O5PS
Quantity 1 mg
Synonym sp-camps, sp-adenosine 3',5'-cyclic monophosphorothioate triethylammonium salt
InChI Key OXIPZMKSNMRTIV-UHFFFAOYSA-N
SMILES CC[NH+](CC)CC.C1C2C(C(C(O2)N3C=NC4=C3N=CN=C4N)O)OP(=S)(O1)[O-]
IUPAC Name 6-(6-aminopurin-9-yl)-2-oxido-2-sulfanylidene-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol;triethylazanium
Molecular Weight (g/mol) 446.463
PubChem CID 57369926
Formula Weight 446.46
Percent Purity >98%
Chemical Name or Material cAMPS-Sp, triethylammonium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'