Learn More
(+)-Camptothecin, 98%, Thermo Scientific Chemicals
224.12 € - 934.65 €
Chemical Identifiers
CAS | 7689-03-4 |
---|---|
Molecular Formula | C20H16N2O4 |
Molecular Weight (g/mol) | 348.35 |
MDL Number | MFCD00081076 |
InChI Key | VSJKWCGYPAHWDS-FQEVSTJZSA-N |
Synonym | camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecinShow More |
PubChem CID | 24360 |
ChEBI | CHEBI:27656 |
SMILES | CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10502461
|
Thermo Scientific Acros
276721000 |
100 mg | Glass bottle |
224.12 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772315
|
Thermo Scientific Acros
276720010 |
1 g | Glass bottle |
934.65 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7689-03-4 | |
348.35 | |
VSJKWCGYPAHWDS-FQEVSTJZSA-N | |
24360 | |
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O |
C20H16N2O4 | |
MFCD00081076 | |
camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecinShow More | |
CHEBI:27656 |
Specifications
7689-03-4 | |
100.0 | |
Yellow | |
Authentic | |
C20H16N2O4 | |
15, 1737 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetonirile,dmso,,chloroform,hydrochloric acid | |
CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O | |
24360 | |
348.35 | |
Powder |
97.5 | |
260.0°C | |
1.0% max. | |
97.5% min. (HPLC) | |
MFCD00081076 | |
camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin | |
VSJKWCGYPAHWDS-FQEVSTJZSA-N | |
348.35 | |
CHEBI:27656 | |
98% | |
(+)-Camptothecin, 98% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : UQ0492000
missing translation for 'tsca' : TSCA
- (+)-Camptothecin
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.