missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytotoxic plant alkaloid with antitumor properties; prototypic DNA topoisomerase I inhibitor. Induces single strand DNA breaks and protein-DNA crosslinks.

Specifications
Specifications
CAS | 7689-03-4 |
Molecular Formula | C20H16N2O4 |
Quantity | 25 mg |
Synonym | camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin |
InChI Key | VSJKWCGYPAHWDS-FQEVSTJZSA-N |
SMILES | CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O |
Molecular Weight (g/mol) | 348.36 |
PubChem CID | 24360 |
ChEBI | CHEBI:27656 |
Formula Weight | 348.36 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Camptothecin, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction