missing translation for 'onlineSavingsMsg'
Learn More

Manutan Can, 1.0L open top metal lacquered interior with lid

Brand:  Manutan A350777

Encompass_Preferred

Product Code. 12846149

  • 62.80 € / Pack of 24

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Suitable for storing paints, coatings, adhesives and chemicals. All containers have welded side seams and re-sealable lids. Plain or lacquered tinplate interior.
  • Capacity: 1L
  • Dimensions (H x D): 120mm x 113mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers