missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (-)-Cannabidiol

Natural cannabinoid; GPR55 antagonist, weak CB1 antagonist, CB2 inverse agonist and AMT inhibitor

240.00 € - 904.00 €

Specifications

Chemical Name or Material 2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol
CAS 13956-29-1
Target Non-selective Cannabinoid Receptor Antagonists
Molecular Formula C21H30O2
Purity 0.99
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18094992
View Documents
Tocris Bioscience™
1570/10
10 mg
240.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18075332
View Documents
Tocris Bioscience™
1570/50
50 mg
904.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(-)-Cannabidiol is a non-psychotropic constituent of cannabis that is anticonvulsive, antihyperalgesic and neuroprotective in vivo. GPR55 and weak CB1 antagonist (IC50 values are 0.445 and 3.35 μM), CB2 receptor inverse agonist and inhibitor of anandamide uptake (IC50 = 27.5 μM). Also inhibits FABPs and a weak agonist at TRP vanilloid receptors (EC50 = 3.5 μM).
Specifications

Specifications

2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentyl-1,3-benzenediol
Non-selective Cannabinoid Receptor Antagonists
0.99
13956-29-1
C21H30O2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications