Learn More
Canrenone, MedChemExpress
MedChemExpress Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent.
Brand: Medchem Express HY-B1438
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent.Specifications
Canrenone | |
Light Yellow | |
C22H28O3 | |
Aldadiene SC9376 | |
C[C@@]12[C@]3(CCC(O3)=O)CC[C@@]1([H])[C@]4([H])C=CC5=CC(CC[C@]5(C)[C@@]4([H])CC2)=O | |
340.46 | |
Research | |
COVID-19-immunoregulation |
976-71-6 | |
Research | |
50 mg | |
DMSO : ≥ 100 mg/mL (293.72 mM) | |
340.46 | |
99.54% | |
Powder |