Learn More
Hirschmann™ Dispsable Capillary Tubes
Excellent for working with small samples in the laboratory. Sodium-heparinized disposable blood gas analysis capillary pipettes.
Brand:  Hirschmann™ 9221275
Product Code. 10049640
  • 161.00 € / Pack of 1000
Explore more special offers
Description
Description
  • 225 i. u. Sodium
  • e.g. for blood gas analysis
  • CE acc. IVD 98/79EC
Specifications
Specifications
Capillary Tube
100 μL
1.35 mm
75 mm