Learn More
Capillary column 123-1033 DB-1 fused silica 1μm film thickness 30m x 0.32mm J and W
Brand:  Agilent Technologies 123-1033
Encompass_Preferred
Product Code. 13118113
  • 1152.95 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Capillary column 123-1033 DB-1 fused silica 1μm film thickness 30m x 0.32mm J and W