Learn More
Jaytec Glass™ Capillary Sealing Compound
Capillary Sealing Compound is used to plug the end of a capillary tube. The kit consists of a non-drying cement in a plastic tray. Push the empty end of a filled capillary tube into the paste to seal it.
Brand: Jaytec Glass™ CAP-CS-10
This item is not returnable.
View return policy
Description
- 24 capillaries can be placed upright in the numbered storage holes on either side of the plate.
Specifications
Plastic | |
Sealing Compound |
Product Content Correction
Product Title
Jaytec Glass™ Capillary Sealing Compound > Capillary Sealing Compound
Spot an opportunity for improvement?Share a Content Correction