Learn More
IKA™ Capillary Set
The capillary is used to regulate the flow rate range. It is determined by the size of the capillary and viscosity of the solvent.
Brand:  IKA™ 0020103091
Product Code. 17563636
  • 328.00 € / Pack of 4
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For regulating the flow rate range.
  • It is determined by the size of the capillary and viscosity of the solvent.
  • The set contains capillary tubes with four different sizes.
  • Scope of delivery
    • Capillary assembly/125 μm/50 mm
    • Capillary assembly/125 μm/100 mm
    • Capillary assembly/180 μm/50 mm
    • Capillary assembly/250 μm/100 mm
Specifications
Specifications
Four Different Sizes