Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
Brand:  Hirschmann™ 9211575
 View more versions of this product
Product Code. 10587441
  • 82.75 € / Pack of 1000
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Specifications
1.15 mm
Melting point determination
75 mm
1.55 mm
One end closed
Capillary Tube