Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
Brand:  Hirschmann™ 9211790
 View more versions of this product
Product Code. 10652812
  • 130.00 € / Pack of 1000
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Specifications
1.3 mm
Melting point determination
90 mm
1.75 mm
One end closed
Capillary Tube